Learn More
Nickel Electro™ Stainless Steel Funnels
This durable stainless steel funnel, break-resistant, stain proof and easy to clean makes it an ideal choice for most laboratory and research environments.
Brand: Nickel Electro™ FUNSS080H
5.54 GBP valid until 2024-10-18
Use promo code "21642" to get your promotional price.
Description
- Laboratory Funnel, with handle, stainless steel 304 spec 18/8 non-magnetic, bright finish
- Durable stainless steel funnel, break-resistant, stain proof and easy to clean
- May be used with or without filter membranes for gravity filtration as well as simple funnel procedures
- Flute is vented to allow complete, uninhibited flow
- Can be used with liquids or solids
- The handle can also be used as a convenient hanging point when not in use
- Smooth finish on internal surfaces making funnel easy to clean or sterilise (sterilize)
Specifications
Stainless Steel | |
Vented flute |