Learn More
SPEX™ Stainless Steel Grinding Ball
SPEX™ Stainless Steel Grinding Ball is a replacement ball for use with 3114 Stainless Steel Grinding Vial Set.
Brand: SPEX™ 3114SB
Code : NEW
Additional Details : Weight : 5.00000kg
Description
- Durable and tough grinding ball suitable for general purpose grinding
- High durability and efficiency with moderate resistance to abrasion
- Resists chemical attack
- Typical composition: Major Elements – Fe, Cr; Minor Elements – Ni, Mn, S, Si
- Hardness: Mohs 5-5 1/2; Rockwell – C 55-60
Specifications
6.35 mm | |
3114 Stainless Steel Grinding Vial Set | |
Stainless Steel | |
4 Balls |