missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Grinding Balls for Geno/Grinder™ Homogenizers

Use grinding balls to grind samples in Geno/Grinder™ homogenizers.

Brand:  Cole-Parmer™ 2156

Code : NEW

Additional Details : Weight : 3.00000kg

 View more versions of this product

Product Code. 13591160

  • £392.00 / Pack of 100
Estimated Shipment: 24-09-2024
to see stock.
Add to basket

Description

Description

  • Suitable for use with the Cole-Parmer™ Geno/Grinder™ homogenizers.
  • Optimize performance of your Geno/Grinder™ homogenizers with a wide range of grinding ball options suitable for your application.
Specifications

Specifications

Grinding Balls
1.1 cm
24-Well Titer Plates, 15 mL Short Polycarbonate Vials of Geno/Grinder™
0.43 in.
Ideal for Grinding Large or Tough Samples
Stainless Steel (440 C)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers