missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Stainless Steel Grinding Balls

Designed for use with Retsch™ mixer mills. Retsch™ Stainless Steel Grinding Balls are ideal for disruption of biological cells as well as for DNA/RNA extraction.

Brand:  RETSCH 22.455.0002

Code : 50

Additional Details : Weight : 0.02250kg

 View more versions of this product

Product Code. 11758414

  • £97.00 / Pack of 200
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • For use with MM 200 and MM 400 Mixer Mills
Specifications

Specifications

Grinding Balls
3 mm
Reaction Vials
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers