Learn More
RETSCH Stainless Steel Grinding Balls
Designed for use with Retsch™ mixer mills. Retsch™ Stainless Steel Grinding Balls are ideal for disruption of biological cells as well as for DNA/RNA extraction.
£5.80 - £242.00
Product Code | Brand | Diameter (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||||
10208591
|
RETSCH
05.368.0060 |
40 mm | Planetary Ball Mill PM 400 |
£71.45
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
|||||
10729954
|
RETSCH
05.368.0109 |
15 mm | MM 200 and MM 301 |
£15.00
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10146641
|
RETSCH
22.455.0003 |
5 mm | Reaction Vials |
£242.00
Pack of 200 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10054460
|
RETSCH
05.368.0035 |
7 mm | MM 200 and MM 301 |
£5.80
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
10539711
|
RETSCH
05.368.0034 |
5 mm | MM 200 and MM 301 | N/A | ||||||
11758414
|
RETSCH
22.455.0002 |
3 mm | Reaction Vials |
£99.70
Pack of 200 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
10196541
|
RETSCH
22.455.0002 |
3 mm | Reaction Vials |
£101.00
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
|||||
11727664
|
RETSCH
224550001 |
4 mm | Reaction Vials |
£143.00
Pack of 200 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
Description
- For use with MM 200 and MM 400 Mixer Mills
Specifications
Grinding Balls | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
RETSCH Stainless Steel Grinding Balls