missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Stainless Steel In-Line Filter Holder

Brand:  Cole-Parmer™ 02929-20

Code : A0

Additional Details : Weight : 0.58000kg

Product Code. 11791953

  • £621.80 / Each
Estimated Shipment: 24-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Autoclavable
Stainless Steel
57 mm
In-line Filter
1 Each
69 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers