Learn More
Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
£213.00
Product Code | Brand | Model | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | For Use With (Equipment) | Price | Quantity & Availability | ||||
10458732
|
Bandelin Electronic™
359 |
K10 | Ultrasonic Bath, RK/DT 510 / 510 H / 510 H-RC, DL 510 H |
£213.00
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||||
Description
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
- Available for a wide range of tank capacities
- Available in a range of mesh sizes
- Excellent for the cleaning of technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Basket Insert | |
Stainless Steel |