missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, A286, S66286, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Stainless Steel, A286, S66286, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Nitrogen/Tin/Boron/Niobium/Phosphorus/Tungsten/Carbon/Cobalt/Copper/Silicon/Vanadium/Manganese/Aluminium/Molybdenum/Titanium/Chromium/Calcium/Antimony/Magnesium/Nickel/Cerium/Zinc/Lead/Selenium/Oxygen/Zirconium/Arsenic |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Nitrogen 0.0035 %/Tin 0.0039 %/Boron 0.0063 %/Niobium 0.007 %/Phosphorus 0.013 %/Tungsten 0.036 %/Carbon 0.038 %/Cobalt 0.04 %/Copper 0.047 %/Silicon 0.05 %/Vanadium 0.223 %/Manganese 0.224 %/Aluminium 0.29 %/Molybdenum 1.23 %/Titanium 2.17 %/Chromium 14.29 %/Calcium (<0.0005) %/Antimony (<0.004) %/Magnesium (<0.005) %/Nickel (<0.008) %/Cerium (<0.015) %/Zinc (<0.02) %/Lead (<0.03) %/Selenium (<0.03) %/Oxygen (0.001) %/Zirconium (0.0016) %/Arsenic (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction