Learn More
Welch Ilmvac™ Stainless Steel KF Reducing Adapter
Designed for use with Welch Ilmvac™ Rotary Vane Pumps.
Brand:  Welch Ilmvac™ 701401
 View more versions of this product
Product Code. 10657852
  • £74.05 / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless Steel KF Reducing Adapter, DN16/25
  • Connect KF 25 for intake or exhaust with CRVpro
Specifications
Specifications
Stainless Steel
1 Each
DN 25/16
KF Reducing Adapter