Learn More
Nickel Electro™ Stainless Steel Beakers with Spout
Will not shatter or chip, this durable stainless steel laboratory beaker is suitabale for laboratory applications.
Brand: Nickel Electro™ 203
Description
- Durable stainless steel laboratory beaker
- 304 spec stainless steel beakers
- Will not shatter or chip
- Suitable for laboratory applications since
- All surfaces are non-reacting
- Offer zero absorption
Specifications
77 mm | |
Stainless Steel | |
Spout |
550 mL | |
124 mm | |
Beaker |