missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Stainless Steel Laboratory Tray
Manufactured by Bochem™, the Stainless Steel Laboratory Tray is useful for containing and carrying items used for experimentation in the lab.
Brand: Bochem™ 8701
This item is not returnable.
View return policy
Description
- Stainless Steel
- Round Angles
- Steel sides to protect contents
Specifications
290 x 220 mm | |
60 mm | |
Stainless Steel | |
180, 220 mm | |
Rectangular |
290 x 220 x 60 mm | |
250, 290 mm | |
Laboratory Tray | |
1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction