Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Product Code. p-8004963
Click to view available options
Needle Gauge:
22 G
22s G
23 G
23s G
25s G
26 G
26s G
Needle Point Style:
2, 3, or 4
5 or AS
Unit Size:
Each
Pack of 6
Product Code. 10146383
Brand: Hamilton™ 773201
Can be used with removable (RN)-style syringes that are 250μL and larger.
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style and lengths
  • Autoclavable
  • For large hubs
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Needle Gauge 22 G
Diameter (Metric) Inner 0.41 mm
Diameter (Metric) Outer 0.72 mm
Product Type Removable Custom Needle
Quantity 6/Pk.
Needle Point Style 5 or AS
For Use With (Equipment) Removable Syringes (250μL and larger)