Learn More
Cole-Parmer™ Stainless Steel Luer Fittings
Exceptionally durable Luer fittings suitable for harsh chemical environments. Cole-Parmer™ Stainless Steel Luer Fittings are easy to disassemble for cleaning.
Brand:  Cole-Parmer™ 41507-32
 View more versions of this product
Product Code. 15234743
  • £121.00 / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Exceptionally durable Luer fittings suitable for harsh chemical environments. Cole-Parmer™ Stainless Steel Luer Fittings are easy to disassemble for cleaning.
  • Exceptional durability
  • Easy to disassemble for cleaning
  • Ideal for harsh chemical environments
  • Autoclavable
Specifications
Specifications
Autoclavable
Harsh chemical environments
Male Luer lock x 1/4-28 UNF thread
Stainless Steel
1 Pack
Luer Fitting
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Stainless Steel Luer Fittings > Includes: Male Luer lock x 1/4-28 UNF thread