Learn More
Hamilton™ Stainless Steel Luer Lock Metal Hub Needle, Custom length, Point Style 2, 3, or 4
Metal hub makes the most rigid connection to the needle making it perfect for repetitive septum piercing.
Brand:  Hamilton™ 7749-05
 View more versions of this product
Product Code. 16522338
  • £220.00 / Each
Edge
Explore more special offers
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Available in various gauges
  • Autoclavable
  • Metal hubs manufactured from nickel plated brass
  • Custom length
  • Compatible with PTFE Luer Lock (TLL) syringes
  • Design controlled by a series of universal standards which guarantees compatibility between manufacturers
  • Provided in packs of 6
Specifications
Specifications
Autoclavable
14 G
2.11 mm
6/Pk.
TLL Syringes and LT or TLL Connectors
Nickel-plated Brass (Hub), Stainless Steel
1.16 mm
Luer Lock Custom Needle
2, 3, or 4
Standard Tapered