Learn More
Fisherbrand™ Stainless Steel Manual Crimper for clean room, adjustable
Provides a reproducible, secure vial closure
Brand:  Fisherbrand™ 20 06 2229
831.25 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11757746
  • £889.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless Steel
  • Sterilizable for cleanroom applications
  • Manual
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20mm Crimper
20 mm
Stainless Steel