Learn More
Contec™ Stainless Steel Multi-Bucket System
GREENER_CHOICEBIOPHARMA
Stainless Steel Multi-Bucket System offers maximum flexibility and can be used with two or three 7 gallon (27 L) buckets depending upon your in-house protocols.
Brand:  Contec™ 2707
1883.36 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16681892
  • £1985.00 / Each
Estimated Shipment: 28-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Lightweight, stainless steel, and easy to maneuver multi-bucket system offers maximum flexibility and can be used with two or three 7- gallon (27 L) buckets depending upon in-house protocols.
  • An extendable frame pulls out of the front of the fully autoclavable cart to accommodate an additional bucket, part number 2617.
  • The cart features non-marring run quiet wheels.
  • Buckets are deep drawn and seamless reducing the risk of microbial contamination
  • The removable handle makes it convenient for autoclaving and storage.
  • A lip around the top edge of the buckets allows for easy lifting.
  • Autoclavable at 121°C for 30 minutes
  • 2700 multi-bucket system includes heavy-duty stainless steel downpress wringer that is compatible with all sizes of VertiKlean™, VertiKlean™ MAX™, QuickTask™, and Edgeless™ Mops.
  • 2707 multi-bucket system offers the ability to customize the system with whichever wringer best coincides with in-house protocols. The system is compatible with two stainless steel downpress wringer options to accommodate both flat and string mops
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Multiple Bucket System
Autoclavable
Ideal for use in life sciences, microelectronics, semiconductor or any critical manufacturing environment
Non-sterile
Stainless Steel
Reusable
Two 7 gal. Buckets, cart with casters (wringer options sold separately)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Contec™ Stainless Steel Multi-Bucket System > Includes: Two 7 gal. buckets, cart with casters