Learn More
Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight Sets Without DAkkS Calibration Certificate
Brand:  Sartorius™ YCS011-511-00
3750.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10439311
  • £3885.00 / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Wood
1 mg to 100 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius™ Stainless Steel OIML Class E1 Reference Line Weight Sets Without DAkkS Calibration Certificate > Mass: 1mg to 100g