missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Stainless Steel PharmaScoop™
Constructed of high quality Stainless Steel for durability. Sampling Systems™ Stainless Steel PharmaScoop™ is ideal for dispensing and sampling in pharmaceutical and hygienic environments.
Brand: Sampling Systems™ A316-10
This item is not returnable.
View return policy
Description
- Very high quality scoops
- Heavy duty construction
- Made from 316L Stainless Steel
- Crevice free
- Satin finish
- Rounded corners for easy cleaning
- GMP correct
Specifications
Powder Scoop | |
10 mL | |
25 mm | |
45 mm |
Stainless Steel (316L) | |
Mirror Polish | |
75 mm |