Learn More
Buerkle™ Stainless Steel PharmaSpoon™
Buerkle™ Stainless Steel PharmaSpoon is designed to take samples in the pharmaceutical industry.
Brand:  Buerkle™ 5324-0001
Product Code. 10196514
  • £727.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For taking samples of powdery substances, intermediate products and material in the pharmaceutical industry
  • Special materials with excellent surface qualities must be used for this to prevent cross-contamination
  • Made of stainless steel AISI 316L (1.4404)
  • Surface highly polished by hand (not electro-polished)
  • Surface properties of the PharmaSpoon provide significant advantages
  • Possible residues are even visible in the trace sector (μg sector)
  • The surface is easy to clean
  • The PharmaSpoon is also suitable for sterile sampling
Specifications
Specifications
Spoon
Autoclavable
Foodstuffs
Stainless Steel (V4A)
High polish
650 mm