Learn More
Stainless Steel powder, -20+35 mesh, Type 316-L, Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 043460.A3
Code : NEW
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 11345777
  • £169.00 / 2kg
In Stock
to see stock.
Description
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Stainless Steel powder, Type 316-L
MFCD00148499
Insoluble in water.
Powder
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
2 kg
−20+35 Mesh
Product Suggestions
Product Suggestions
Videos
SDS Documents