missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel powder, -40+80 mesh, Type 316, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 042944.A1

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11310218

  • £180.00 / 1kg
Estimated Shipment: 19-08-2024
to see stock.

Description

Description

Stainless Steel powder, Type 316 is a increased resistance to chemical corrosion for the addition of Molybdenum. Also used in a wide variety of home and commercial applications, including, chemical process equipment, food and beverage industry and fasteners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Stainless Steel powder, Type 316
MFCD00148499
Insoluble in water.
Powder
Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
1 kg
−40+80 Mesh
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only