missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Stainless Steel Propellers

Use this four-blade propeller for general-purpose vortex mixing. Cole-Parmer™ Stainless Steel Propellers are ideal for a variety of applications.

£73.29 - £105.00

Specifications

Quantity 1 Each
Type Propeller
View More Specs

Products 6
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
11754639
View accessory products View Documents Promotion Details
Cole-Parmer™
04370-13
40 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11714739
View accessory products View Documents Promotion Details
Cole-Parmer™
04349-10
50.8 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11729998
View accessory products View Documents Promotion Details
Cole-Parmer™
04370-19
28 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11724739
View Documents Promotion Details
Cole-Parmer™
04349-00
774.7 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11739998
View accessory products View Documents Promotion Details
Cole-Parmer™
04370-11
75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11749998
View accessory products View Documents Promotion Details
Cole-Parmer™
04370-12
60 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Stainless steel four-blade propeller
  • Threaded 10 mm dia. bore 
  • Fits only shaft model 04370-02
Specifications

Specifications

1 Each
Propeller
Videos
SDS
Documents

Documents

Product Certifications