missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Stainless Steel Quali Rod Cylindrical

Cylindrical hollow drill with a sharp cutting edge, can easily be driven into paste-like and semi-solid substances. The samples cut out are subsequently removed with the appropriate push-rod ejector. Total length 260 mm. Inner Ø 8 mm, outer Ø 10 mm.

Brand:  Buerkle™ 5311-0101

145.73 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


Product Code. 10260082

  • £165.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Cylindrical hollow drill with a sharp cutting edge, can easily be driven into paste-like and semi-solid substances. The samples cut out are subsequently removed with the appropriate push-rod ejector. Total length 260mm. Inner diameter 8mm, outer diameter 10mm.
  • Stainless steel V2A (1.4301)
  • Sterilizable
  • Inner diameter 8mm, outer diameter 10mm
Specifications

Specifications

Quali Rod Cylindrical
Stainless Steel
260 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.