Learn More
Grant Instruments™ Stainless Steel Perforated Base Tray
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
Brand:  Grant Instruments™ SBT6
Code : 57
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11406417
  • £99.11 / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
  • For use with SBB Aqua Plus and LSB Series Aqua Pro Series Unstirred Water Bath
  • Choose from a vartiety of sizes.
Specifications
Specifications
DMU5, SAP2/5, JBN5
SBT6 Base Tray