Learn More
Grant Instruments™ Stainless Steel Perforated Base Tray
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
Brand: Grant Instruments™ SBT2
Code : 57
Additional Details : Weight : 0.10000kg
Description
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
- Base tray stainless steel for use with Sub Aqua Pro 2L
Specifications
SAP2 | |
Perforated Base Tray | |
Stainless Steel |