Learn More
Grant Instruments™ Stainless Steel Perforated Base Tray
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
Brand: Grant Instruments™ SBT14
Code : 57
Additional Details : Weight : 0.10000kg
Description
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
- For use with SBB Aqua Plus and LSB Series Aqua Pro Series Unstirred Water Bath
- Choose from a vartiety of sizes.
Specifications
DMU12 & SAP12 | |
SBT14 Stainless Steel Base Tray |