missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Stainless Steel Safety Canister
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
£173.84 - £237.44
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
17826476
|
asecos™
40090 |
5 L | 169.00 mm |
£173.84
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
17876536
|
asecos™
36484 |
10 L | 280.00 mm |
£208.82
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
17816406
|
asecos™
40092 |
20 L | 271.00 mm |
£220.48
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
17846486
|
asecos™
36485 |
20 L | 470.00 mm |
£237.44
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
17806506
|
asecos™
40091 |
10 L | 241.00 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Incl. multilingual safety sticker for content labelling, with filling tap made of stainless steel (1.4301)
- Incl. ventilation, NBR seals and aluminium screw cap integrated flame arrester and plastic protective ring
Specifications
Gray | |
Stainless Steel |