Learn More
Fisherbrand™ Stainless Steel Sharp Ends Forceps
Product Code. p-8013182 Shop All Fisherbrand Products
Click to view available options
Height (Metric) Body:
115 mm
130 mm
160 mm
Unit Size:
Each
Product Code. 11562113
Only null left
Add to basket
Edge
Add to basket
Provides a non-stick surface with PTFE coating to which very few solid or liquid substances will permanently adhere. Nickel Electro™ Stainless Steel Sharp Ends Forceps provide excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized.
PTFE coated forceps, providing a non-stick surface to which very few solid or liquid substances will permanently adhere.
  • Chemically resistant and non-contaminating
  • Non-protein binding and providing bio-inert surfaces
  • No critical quantities lost due to sticking
  • No build up on surfaces
  • Ideal for use under severe laboratory conditions
  • Excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized
  • With guide pin
Specifications
Tip Style Sharp
No. per Pack Each
Material PTFE-coated Stainless Steel
Product Type Forceps
Height (Metric) Body 115 mm
Style Serrated Jaws