missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh
Made from stainless steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has an 100 mm diameter.
Marke: Fisherbrand™ 01/100SBW250um
Beschreibung
100 mm diameter test sieves with stainless steel frame and woven wire mesh. Supplied with Certificate of Conformity and record card. Additional mesh sizes and diameter sieves available - please contact Customer Service for details.Spezifikation
1 Ea. | |
Sieve | |
Stainless Steel | |
100 mm | |
Woven Wire |
250 μm | |
Part 1 : 2000, ISO 3310 : Part 1 : 2000, BS410 | |
Certificate of conformity and record card | |
Stainless Steel |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh > Pore Size: 250 μm
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur