Learn More
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill
Depending on the sample, even finer results can be achieved by grinding with two or more sieves with progressively smaller hole sizes.
£137.00 - £167.00
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10012393
|
Kinematica™
35095008 |
0.8 mm |
£153.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
10387183
|
Kinematica™
35095030 |
3 mm |
£137.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10713454
|
Kinematica™
35095050 |
5 mm |
£144.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10754024
|
Kinematica™
35095060 |
6 mm |
£144.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10042293
|
Kinematica™
35095002 |
0.2 mm |
£167.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10275893
|
Kinematica™
35095005 |
0.5 mm |
£144.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10532274
|
Kinematica™
35095010 |
1 mm |
£162.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10134133
|
Kinematica™
35095015 |
1.5 mm |
£162.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10617693
|
Kinematica™
35095020 |
2.0 mm |
£154.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10677123
|
Kinematica™
35095040 |
4 mm |
£144.00
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
Specifications
Sieve | |
Stainless Steel |