Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for IF750, Reinforced Chamber Option
Non-perforated slide-in drip trays constructed of stainless steel.
Brand: Memmert™ IF750B32763
Additional Details : Weight : 0.01200kg
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
IF750 Incubator | |
Stainless Steel | |
Drip Tray | |
Rim: 15 mm |