Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN75plus, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Brand:  Memmert™ SN75plusE02072
Product Code. 17548785
  • £77.65 / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
Specifications
SN75plus Sterilizer
Drip Tray
Rim: 15 mm
Stainless Steel