Learn More
Memmert™ Stainless Steel Slide-In Drip Tray
Stainless steel slide-in bottom drip tray
Brand:  Memmert™ HPP260ecoE29726
 View more versions of this product
Product Code. 17129784
  • £172.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • May affect the temperature distribution
  • 15 mm rim
Specifications
Specifications
Stainless steel slide-in drip tray, 15 mm rim (may affect the temperature distribution) - cannot be used in connection with option K1
HPP260eco
15 mm