Learn More
Memmert™ Stainless Steel Slide-In Drip Tray
Stainless steel slide-in bottom drip tray
Brand:  Memmert™ HPP750ecoE02075
 View more versions of this product
Product Code. 17129874
  • £238.36 / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • May affect the temperature distribution
  • 15 mm rim
Specifications
Specifications
Stainless steel slide-in drip tray, 15 mm rim (may affect the temperature distribution) - cannot be used in connection with option K1
HPP750eco
15 mm