Learn More
Memmert™ Stainless Steel Slide-In Drip Tray
Stainless steel slide-in bottom drip tray
Brand:  Memmert™ IPP750ecoB32763
 View more versions of this product
Product Code. 17140255
  • £266.00 / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • May affect the temperature distribution
  • 15 mm rim
Specifications
Specifications
Stainless steel slide-in drip tray, 15 mm rim, with guide bars and fixing screws (may affect the temperature distribution) - can be used only in connection with option K1
IPP750eco
Reinfor Ced Chamber Option
15 mm