Learn More
Memmert™ Stainless Steel Slide-In Drip Tray
Stainless steel slide-in bottom drip tray
Brand:  Memmert™ HPP750ecoB32763
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17189854
  • £238.36 / Each
Estimated Shipment: 16-10-2024
to see stock.
Add to basket
Description
Description
  • May affect the temperature distribution
  • 15 mm rim
Specifications
Specifications
Stainless steel slide-in drip tray, 15 mm rim, with guide bars and fixing screws (may affect the temperature distribution) - can be used only in connection with option K1
HPP750eco
Reinfor Ced Chamber Option
15 mm