Learn More
Memmert™ Stainless Steel Slide-In Drip Tray
Stainless steel slide-in bottom drip tray
Brand:  Memmert™ IPP110ecoE02073
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17190095
  • £111.00 / Each
Estimated Shipment: 16-10-2024
to see stock.
Add to basket
Description
Description
  • May affect the temperature distribution
  • 15 mm rim
Specifications
Specifications
Stainless steel slide-in drip tray, 15 mm rim (may affect the temperature distribution) - cannot be used in connection with option K1
IPP110eco
15 mm