Learn More
Grant Instruments™ Stainless Steel Sloping Lids
Recommended for use above 60°C. Stainless Steel Sloping Lid avoids contamination, reduces evaporation and saves energy.
Brand:  Grant Instruments™ LU6
 View more versions of this product
Product Code. 11416427
  • £192.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Sub Aqua Pro Series Unstirred Water Bath
  • Triangular-shaped stainless steel with plastic handle
  • Choose from the following available sizes:
    • 5L (LU6)
    • 12L (LU14)
    • 18 or 26L (LU28)
    • 34L (LU36)
Specifications
Specifications
SAP5, ST5
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Stainless Steel Sloping Lids > For Use With: SAP5