missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Stainless Steel Spatula
Universal instrument for collecting, dividing and mixing pastes, creams etc.
Brand: Buerkle™ 5313-0180
15.95 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Universal instrument for collecting, dividing and mixing pastes, creams etc.- Blade made of stainless steel V2A (1.4301)
- Wooden handle
Specifications
Spatula | |
Stainless Steel V2A | |
Stainless Steel | |
180 mm |
Universal Instrument for Collecting, Dividing and Mixing Pastes, Creams | |
Wood | |
Wood |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction