missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Spoon/Spatula Combo Utensils

Use one tool for two jobs. Nickel Electro™ Stainless Steel Spoon/Spatula Combo Utensils measure liquids and powders with the spoon end and divide and separate materials with the spatula end.

Brand:  Fisherbrand™ 2013

 View more versions of this product

Product Code. 11864233

  • £54.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Dual-use utensil measures liquids and fine powders with the spoon end; divides and separates materials with the spatula end.
  • Manufactured from stainless steel.
Specifications

Specifications

10 Pk.
Autoclavable
Spatula
Sterile Applications
210 x 20 mm
Stainless Steel
Sterile
2 mL
Bright Finish
20 mm
-196°C to +550°C
210 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.