Learn More
Stainless Steel Spheres, 4mm (0.16 in.) dia., Thermo Scientific Chemicals
£18.20 - £53.80
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355959
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042021.KN
100 Pc.
£53.80
Pack of 100
View alternative products
Save Up to 
»
In Stock
to see stock.
11345959
View Documents Promotion Details
Thermo Scientific Alfa Aesar
042021.KM
25 Pc. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Videos
SDS
Documentos
Documentos