Learn More
S Murray and Co™ Stainless Steel Spring Forceps
Product Code. 12371369
Click to view available options
Length (Metric):
115 mm
125 mm
150 mm
Tip Style:
Blunt, Narrow Flutes
Sharp, Curved, Narrow Flutes
Sharp, Narrow Flutes
Unit Size:
Each
Product Code. 12371369
Brand: S Murray and Co™ E004/02
Only null left
Add to basket
Edge
Add to basket
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
  • Stainless Steel forceps for dissection
Specifications
Product Type Spring Forceps
Material Stainless Steel
For Use With (Application) For dissection
Quantity Each
Tip Style Sharp, Narrow Flutes
Length (Metric) 125 mm