Learn More
S Murray™ Stainless Steel Spring Forceps
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
Brand:  S Murray™ E006/02
 View more versions of this product
Product Code. 12381369
  • £17.30 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
  • Stainless Steel forceps for dissection
Specifications
Specifications
Spring Forceps
For dissection
Sharp, Curved, Narrow Flutes
Stainless Steel
Each
125 mm