Learn More
Bochem™ Stainless Steel Stand Base
Support your supplies with the Bochem™ Stainless Steel Stand Base, which has a reinforcing leg for stability.
Brand:  Bochem™ 5002
 View more versions of this product
Product Code. 10363621
  • £117.00 / Each
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Expanded, flat and heavy feet for stability  
  • 1 adjustable foot
  • Anti-slide rubber protection
  • Thread M10
Specifications
Specifications
Stainless Steel (18/10)
300 mm
Stand Base
3 mm
150 mm
1 Pk.