Learn More
Aalborg™ Precision Valve for Stainless Steel Flowmeter
Aalborg™ Precision Valve for Stainless Steel Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
Specifications
For Use With (Equipment) | 150 mm Gas/Liquid Flowmeter |
---|---|
Type | Standard Valve |
Description
- For high precision
- Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
- They may also be used as shut off valves.
- Bodies of the valves are reinforced by structurally rigid metallic shells.
- Shells are made of stainless steel, bushings are made of plated brass or 316 stainless steel.
Specifications
150 mm Gas/Liquid Flowmeter | |
Standard Valve |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Aalborg™ Precision Valve for Stainless Steel Flowmeter