missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Stainless Steel Stirring Paddles

Constructed of high quality stainless steel for durability. Sampling Systems™ Stainless Steel Stirring Paddles are ideal for use in Pharmaceutical and Hygienic applications.

Brand:  Sampling Systems™ A344-1500

 View more versions of this product

Product Code. 15839641

  • £119.00 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Crevice free construction
  • 316L Stainless Steel
  • Satin finish
  • Robust construction
  • BSE/TSE free
Specifications

Specifications

Paddle
Solid Blade
1500 mm
150 mm
Stainless Steel (316L)
Polished
300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers