Learn More
Fisherbrand™ Stainless Steel Stirring Shafts
For use with overhead stirrer units. Fisherbrand™ Stainless Steel Shafts help avoid spillages and ensure even, thorough stirring.
Brand:  Fisherbrand™ A00001311
 View more versions of this product
Product Code. 12396667
  • £55.45 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available with either anchor or 6-hole paddle blades
  • Economically priced
  • Optimum chemical resistance
Specifications
Specifications
Overhead Stirrer
1 Each
0.01 kg
Anchor blade
Stirring Shaft
Stainless Steel