Learn More
asecos™ Stainless Steel Storage Can with Dosing Tap
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
Brand: asecos™ 40084
This item is not returnable.
View return policy
Description
- With fine dosing tap made of stainless steel (1.4301), incl. vent, PTFE seal, integrated flame barrier (removable) and plastic protective ring
- Incl. multilingual safety sticker to designate the contents
Specifications
0.3 gal. | |
Gray | |
5.20 in. | |
8.46 in. | |
1.59 lb. |
1 L | |
Stainless Steel | |
132.00 mm | |
215.00 mm | |
0.72 kg |