Learn More
asecos™ Stainless Steel Storage Can with Screw Cap
The Stainless Steel Safety Canister is ideal for the safe distribution and dosing of flammable liquids.
Marke: asecos™ 40082
Additional Details : Weight : 0.91000kg
Beschreibung
- With screw cap (secured against loss) overpressure valve, PTFE seal, integrated flame barrier (removable) and plastic protective ring
- Incl. multilingual safety sticker to designate the contents
Spezifikation
0.5 gal. | |
Gray | |
6.22 in. | |
8.90 in. | |
2.01 lb. |
2 L | |
Stainless Steel | |
158.00 mm | |
226.00 mm | |
0.91 kg |