Learn More
Socorex™ Stainless Steel Straight Drenchers
Product Code. p-8007532
Click to view available options
Length (Metric):
25 mm
51 mm
102 mm
203 mm
Unit Size:
Pack of 2
Pack of 3
Product Code. 11888432
missing translation for 'mfr': Socorex™ 376.0925S
Use Socorex™ Stainless Steel Straight Drenchers for animal feeding and dosing. These precision-assembled stainless-steel feeding needles are ideal for the delivery of fluid and are reusable since they are autoclavable.
  • Stainless Steel
  • Various lengths to accommodate feeding needs
  • Autoclavable
  • Reusable
Specifications
Autoclavable Autoclavable
Product Type Drenching Cannula
Material Stainless Steel
Length (Metric) 25 mm